Skip to Content
Merck
All Photos(4)

Key Documents

CWA-1186

Sigma-Aldrich

Anti-Human CD98 (MEM-108) ColorWheel® Dye-Ready mAb

greener alternative

Synonym(s):

Large neutral amino acids transporter small subunit 1;4F2 light chain;4F2 LC;4F2LC;CD98 light chain;Integral membrane protein E16;E16;L-type amino acid transporter 1;hLAT1;Solute carrier family 7 member 5;y+ system cationic amino acid transporter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MEM-108, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 55.01 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG1

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

storage temp.

2-8°C

Gene Information

human ... SLC7A5(8140)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone MEM-108 is a mouse monoclonal antibody that detects CD98. It targets an epitope within the extracellular domain.

Immunogen

Burkitt′s lymphoma Raji cell line.

Application

Quality Control Testing

Evaluated by Flow Cytometry in Human peripheral blood mononuclear cells (PBMC) gated on monocytes.

Flow Cytometry Analysis: Staining of one million Human peripheral blood mononuclear cells (PBMC) gated on monocytes was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1186, Anti-Human CD98 (MEM-108) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.

Tested Applications

Flow Cytometry Analysis: Staining of one million Human peripheral blood mononuclear cells (PBMC) gated on monocytes was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1186, Anti-Human CD98 (MEM-108) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Large neutral amino acids transporter small subunit 1 (UniProt: Q01650;also known as 4F2 light chain;4F2 LC;4F2LC;CD98 light chain;Integral membrane protein E16;E16;L-type amino acid transporter 1;hLAT1;Solute carrier family 7 member 5;y+ system cationic amino acid transporter) is encoded by the SLC7A5 (also known as CD98LC;LAT1;MPE16) gene (Gene ID: 8140) in human. CD98 is a multi-pass membrane glycoprotein with seven cytoplasmic domains;twelve transmembrane domains;and six extracellular domains. Its expression is observed in the syncytiotrophoblast layer in the placenta;adult lung;liver;brain;bone marrow;testis;and in resting lymphocytes and monocytes. Lower levels of expression is observed in several other tissues. During gestation;its expression in the placenta is significantly stronger at full-term than at the mid-trimester stage. CD98 has two major biochemical functions. Its heavy chain binds to cytoplasmic tails of integrin-β chains and mediates adhesive si

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 µL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 µL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service