Skip to Content
Merck
All Photos(7)

Key Documents

CWA-1015

Sigma-Aldrich

Anti-Mouse CD45R/B220 (RA3-6B2) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RA3-6B2, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 144.84 kDa

species reactivity

mouse

species reactivity (predicted by homology)

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Ptprc(19264)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone RA3-6B2 is a rat monoclonal antibody that detects Receptor-type tyrosine-protein phosphatase C (CD45R). It targets an epitope within the extracellular domain.

Immunogen

Mouse Abelson Leukemia Virus-Induced pre-B tumor cells

Application

Quality Control Testing:

Evaluated by Flow Cytometry in mouse splenocytes.

Flow Cytometry Analysis (FC): Staining of one million mouse splenocytes was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1015, Anti-Mouse CD45R / B220 (RA3-6B2) ColorWheel® Dye-Ready mAb and Cat. No. CWD-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Rat IgG2a isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Receptor-type tyrosine-protein phosphatase C (UniProt: P06800; also known as EC:3.1.3.48, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5,T200, CD45) is encoded by the Ptprc (also known as Ly-5) gene (Gene ID: 19264) in murine species. CD45 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 26-566), a transmembrane domain (aa 567-588), and a cytoplasmic domain (aa 589-1293). It serves as a receptor-tyrosine phosphatase that is required for antigen-induced signaling in T-cells and B-cells. It contains two protein tyrosine phosphatase (PTPase) domains (aa 642-901 and 933-1216) and the first PTPase domain has enzymatic activity, while the second one seems to affect the substrate specificity of the first one. Its substrate binding site is localized to amino acids 842-848. CD45 functions as a signaling gatekeeper in T-cells that require CD45 to detect and respond to antigen because it dephosphorylates the C-terminal inhibitory tyrosines thereby activates Src family kinase Lck that phosphorylates the T cell antigen receptor (TCR). Upon T-cell activation, CD45 also it recruits and dephosphorylates SKAP1 and Fyn. Furthermore, it also dephosphorylated Lyn and modulates its activity. CD45R is an isoform of CD45 and is expressed at in B cells at all developmental stages from pro-B through mature B cells. It is also detected on abnormal T cells, but not on naïve T cells. Clone RA3-6B2 is reported to be a specific marker for B-cell-type isoform (B220). (Ref.: Courtney, AH., et al. (2019). Sci. Sig. 12(604); eaaw8151; Coughlin, S., et al. (2015). Proc. Natl. Acad. Sci. USA. 112(47); E6515-E6524; Watanabe, Y., and Akaikem T (1994). Scand. J. Immunol. 39(5); 419-425).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service