Skip to Content
Merck
All Photos(1)

Key Documents

CRISPR20V

Sigma-Aldrich

Lenti CRISPR Universal Non-Target Control#3 Transduction Particles (LV04 vector)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51
Pricing and availability is not currently available.

form

liquid

Quality Level

packaging

vial of 200 μL

concentration

(1x106 TU/mL (via p24 titering assay))

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

General description

The Lenti CRISPR Non-Target Control#3 Transduction Particles (LV04 vector) include a gRNA sequence that does not target known human, mouse and rat genes. The non-target control#3 transduction particles are useful as a negative control in experiments using CRISPR lentiviral clones and for cell types that are not amenable to transfection. The Lenti CRISPR Non-Target Control#3 uses a dual-component system consisting of U6-driven guide RNA (non-target#3) and hPGK-driven Puromycin resistance cassette and Blue Fluorescence Protein for fluorophorescence. This control requires Cas9 be delivered by a separate vector.

Lentiviral-based particles permit efficient infection and integration of the construct into differentiated and non-dividing cells, such as neurons and dendritic cells, overcoming low transfection and integration difficulties when using these cell lines. Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging and envelope plasmids.

Application

Functional Genomics/Screening/Target Validation

Other Notes

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells, respectively.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Genome-wide screening with optimized gRNAs per gene ensures specific and efficient knockout, controlling time and cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service