Skip to Content
Merck
All Photos(5)

Key Documents

CMC0008

Sigma-Aldrich

STEADY Chemically Competent Cells

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Product Name

STEADY Chemically Competent Cells, for plasmid production using unstable DNA

biological source

Escherichia coli

grade

for molecular biology

form

buffered aqueous solution

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

competent cell type: chemically competent
transformation efficiency: >1 x 108 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

The STEADY Chemically Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains.
Clones with inverted repeats or other sequnces prone to recombination are commonly found in retroviral genes, and require cells such as this product to be propagated stably.

Genotype

recA13 supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ– leu mtl-1 F– mcrB mrr hsdS20(rB–, mB–)

Features and Benefits

  • Clone unstable sequences with reliable results
  • Available as chemically competent or electrocompetent cells
  • Your choice of two packaging formats
  • High efficiency: over 1 × 108 cfu/μg (chemi) or 1 × 1010 cfu/μg (electro)
  • Excellent value

Components

  • STEADY chemically competent cells
  • pUC 19 transformation control DNA
  • recovery medium for cloning


Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service