Skip to Content
Merck
All Photos(1)

Key Documents

CLS8388

Corning® Costar® traditional, round, plastic storage bottles

round clear polystyrene, sterile, bottle capacity 125 mL, cap (plug seal), case of 24

Synonym(s):

plastic bottle, plastic bottles, storage bottle

Sign Into View Organizational & Contract Pricing

Select a Size

24 EA
$64.30

$64.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
24 EA
$64.30

About This Item

UNSPSC Code:
41121800
NACRES:
NB.14

$64.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

cap (plug seal)
clear polystyrene bottle
cylindrical bottle
red high-density polyethylene cap (plug seal)
round clear polystyrene

sterility

sterile
sterile

feature

cap
graduated
graduations
neck thread wide mouth

packaging

case of 24

manufacturer/tradename

Corning 8388

bottle capacity

125 mL

bottle neck diam.

~44 mm

neck diam.

45 mm

Looking for similar products? Visit Product Comparison Guide

General description

These traditional style disposable polystyrene bottles have smooth sides and are ideal for storage of media, buffers and other aqueous solutions. 45 mm polyethylene plug seal cap provides an airtight seal and helps minimize the risk of contamination. Sterile, certified nonpyrogenic.

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service