Skip to Content
Merck
All Photos(1)

Documents

CLS4854

Corning® gel-loading pipet tips

1-200μL flat 0.4mm thick gel-loading pipet tips, natural, non-sterile, 200 tips/rack, 2 racks/case, 400 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121606
NACRES:
NB.02

material

natural tip
polypropylene

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
packaging: rack

packaging

rack of 200

manufacturer/tradename

Corning 4854

L

83 mm

volume range

1-200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Corning gel-loading polypropylene pipet tips feature a thick capillary end that allows easy access into vertical and horizontal electrophoresis gels.

  • Tips are 83 mm in length
  • Total capacity of 200 μL
  • 200 tips/rack
  • Nonsterile
  • Certified RNase-/DNase-free, nonpyrogenic and trace metal-free

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service