CLS4846
Corning® universal fit pipet tips
1-1000 μL universal fit bulk packed pipet tips, blue, non-sterile, 1000 tips/bag, 10,000 tips/case
Synonym(s):
pipet tips, pipette tips
About This Item
RNase free
barrier: no
disposable
endotoxin free
filter: no
Bulk Pack
case of 10000
Recommended Products
material
blue tip
polypropylene
sterility
non-sterile
feature
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
Bulk Pack
packaging
bag of 1000
case of 10000
manufacturer/tradename
Corning 4846
volume range
1-1000 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning Universal Fit ploypropylene pipet tips are designed to provide a reliable fit with all major brand pipettors and have a bevelled orifice to help ensure fluid delivery. All racked tips are certified RNase/DNase free and nonpyrogenic.
- Reliable fit with all major brand pipettors
- Bevelled orifice to help ensure fluid delivery
- Certified RNase/DNase free and nonpyrogenic
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service