Skip to Content
Merck
All Photos(1)

Key Documents

CLS4806

Corning® universal fit pipet tips

1-200 μL universal fit stack rack pipet tips, natural, non-sterile, 10 racks/case, 4800 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

960 EA
$50.70

$50.70


Estimated to ship onMay 30, 2025



Select a Size

Change View
960 EA
$50.70

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1—200 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation
packaging:
rack of 480
manufacturer/tradename:
Corning 4806

$50.70


Estimated to ship onMay 30, 2025


material

natural tip
polypropylene

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation

packaging

rack of 480

manufacturer/tradename

Corning 4806

volume range

1-200 μL

volume

1—200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Corning® Universal Fit Pipet Tips

Corning Universal Fit ploypropylene pipet tips are designed to provide a reliable fit with all major brand pipettors and have a bevelled orifice to help ensure fluid delivery. All racked tips are certified RNase/DNase free and nonpyrogenic.

  • Reliable fit with all major brand pipettors
  • Bevelled orifice to help ensure fluid delivery
  • Certified RNase/DNase free and nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service