Key Documents
CLS4527
Corning® spheroid microplates
wells (1536), round black/clear bottom, ULA (Ultra-Low Attachment surface), sterile
Synonym(s):
3D cell culture plates
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
material:
polystyrene
round black/clear bottom
size:
1536 wells
Pricing and availability is not currently available.
material
polystyrene
round black/clear bottom
sterility
sterile
feature
ULA (Ultra-Low Attachment surface)
Lid Included
Recommended Medium Well Volume
Recommended Working Volume
Specific plates for 3D cell culture
manufacturer/tradename
Corning 4527
size
1536 wells
well volume
(14 mL)
wells
(1536)
suitability
suitable for (cell cuture)
General description
Key Features:
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel proprietary well geometry aids formation of spheroids in center of well
  • Unique design that shields each well to minimize well-to-well cross talk
Key Benefits:
  • Uniform, single spheroid formation across all wells
  • Centering of the spheroid in each well enables automated visualization
  • Culture and assay spheroids in the same microplate, without the need for transfer to a new plate
  • Ease of media exchange or treatment with test compounds/drugs
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Discover how Corning® Spheroid Microplates played a key role in the use of use of 3D cell cultures for in vitro drug discovery assay.