Key Documents
CLS4510
Corning® 384 well microplate, low volume
High Binding surface treatment, black polystyrene, round bottom, non-sterile, lid: no, pack of 10
Synonym(s):
384 multiwell plates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
material:
black bottom
black polystyrene
round bottom
size:
384 well
sterility:
non-sterile
binding type:
high binding surface
feature:
lid: no
skirt
High Binding surface treatment
Pricing and availability is not currently available.
material
black bottom
black polystyrene
round bottom
sterility
non-sterile
feature
lid: no
skirt
High Binding surface treatment
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 4510
size
384 well
well volume
35 μL
wells
384 wells , low volume
working volume
2-20 μL
binding type
high binding surface
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)