Skip to Content
Merck
All Photos(2)

Key Documents

CLS431489

Corning® Spin-X® UF concentrators

Spin-X UF 20 for samples up to 20mL; 30K MWCO

Synonym(s):

centrifuge columns, desalting columns

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
$197.00

$197.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
12 EA
$197.00

About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

$197.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


material

polycarbonate housing
polyethersulfone membrane

description

white, polypropylene screw cap

sterility

non-sterile

packaging

case of 12 ea

manufacturer/tradename

Corning 431489

H × diam.

116 mm × 30 mm

membrane surface area

6 cm2

tube size

50 mL

working volume

14 mL , fixed angle rotor

pore size

30 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

Corning Spin-X UF Concentrators:
  • Corning® Spin-X® UF centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery
  • The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides fast, high speed concentrating, even with particle laden solutions
  • Integrated dead stop design eliminates the risk of spinning to dryness; no respinning necessary
  • Choice of three sizes for greater flexibility: Spin-X UF 500 for samples up to 500 μL, Spin-X UF 6 for samples up to 6mL, Spin-X UF 20 for samples up to 20 mL (14 mL if using fixed angle rotors)
  • Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5K, 10K, 30K, 50K and 100K to meet all your concentrating needs. Note: Choose a MWCO one-half to one-third smaller than the protein to be concentrated
  • The MWCO and graduations are printed right on the side of the concentrator tube to avoid confusion

Summary:
  • Spin-X UF 500 for samples up to 500 μL
  • Spin-X UF 6 for samples up to 6 mL
  • Spin-X UF 20 for samples up to 20 mL (14 mL if using fixed angle rotors)

Note: Spin-X UF 500 concentrators require fixed angle rotors. Minimum rotor angle is 40º
Note: sample volume of 14mL if using fixed angle rotors

Application

Key Spin-X UF Concentrator Applications:
  • Concentration, desalting of proteins, enzymes, monoclonal antibodies, immunoglobulins
  • Removal of labeled amino acids and nucleotides
  • Deproteinization of samples
  • HPLC sample preparation
  • Recovery of biomolecules from cell culture supernatants, lysates
  • Concentrating virus from cell culture medium

Legal Information

Corning is a registered trademark of Corning, Inc.
SPIN-X is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service