CLS430168
Corning® cell culture flasks
surface area 25 cm2, canted neck, cap (plug seal)
Synonym(s):
culture flask, flask cell culture, tissue culture flask
About This Item
Recommended Products
material
canted neck
cap (plug seal)
plug-seal screw-cap
polystyrene
rectangular bottom flask
rectangular flask
sterility
sterile
feature
cap
graduations
packaging
case of 500
manufacturer/tradename
Corning 430168
maximum volume
10 mL
surface area
25 cm2
working volume
5-7.5 mL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Manufactured from optically clear virgin polystyrene
- Treated for optimal cell attachment
- Printed with lot numbers for ease in traceability
- 100% integrity tested
- Sterilized by gamma irradiation and certified nonpyrogenic
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service