Skip to Content
Merck
All Photos(1)

Key Documents

CLS4138

Corning® DeckWorks barrier pipet tips

1 - 200μL low binding barrier pipet tips, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case

Synonym(s):

low binding barrier pipet tips, low binding barrier pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

960 EA
$457.00
4 X 960 EA
$772.00

$457.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
960 EA
$457.00
4 X 960 EA
$772.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
1—200 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
disposable
endotoxin free
filter
graduated
Tip shape: round
packaging:
pack of 960
manufacturer/tradename:
Corning 4138

$457.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

natural tip
polypropylene

sterility

sterile

feature

DNase free
RNase free
barrier
disposable
endotoxin free
filter
graduated
Tip shape: round

packaging

pack of 960

manufacturer/tradename

Corning 4138

volume range

1-200 μL

volume

1—200 μL

suitability

suitable for (quantitative and semi-quantitative assays such a real-time PCR (qPCR) and conventional PCR)

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Corning® DeckWorks Barrier Tips

Corning DeckWorks barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.

This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.

The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.

Medical grade resins and production excellence allow for full certification as:

  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free
  • Sterility Assurance Level 10-3

Legal Information

Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service