Key Documents
CLS4069
Corning® Lambda® Plus pipettor starter kit
laboratory pipettor starter kit, w/ four Lambda Plus single-channel pipettors conveniently packaged w/ useful accessories, 1/cs
Synonym(s):
laboratory pipettor starter kits
Select a Size
1 EA
$1,430.00
$1,430.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$1,430.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$1,430.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
feature
single-channel
packaging
case of 1
pack of 1
manufacturer/tradename
Corning
Related Categories
General description
Corning® Lambda® Plus Starter kit
The Lambda Plus Starter Kit is designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
The Lambda Plus Starter Kit contains: four Lambda Plus Single-Channel pipettors conveniently packaged with useful accessories (manual, calibration key, color-coded rings, tube of piston grease and identification stickers).
  • CLS4071 Lambda Plus Single-Channel pipettor, 0.5 to 10 μL
  • CLS4072 Lambda Plus Single-Channel pipettor, 2 to 20 μL
  • CLS4074 Lambda Plus Single-Channel pipettor, 20 to 200 μL
  • CLS4075 Lambda Plus Single-Channel pipettor, 100 to 1000 μL
3 Racks of Corning IsoTip® filtered pipet tips:
  • CLS4808, 0.5 μL to 10 μL IsoTips, rack of 96
  • CLS4823, 1 μL to 200 μL IsoTips, rack of 96
  • CLS4809, 100 μL to 1000 μL IsoTips, rack of 96
Legal Information
Corning is a registered trademark of Corning, Inc.
Isotip is a registered trademark of Corning, Inc.
Lambda is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)