Skip to Content
Merck
All Photos(1)

Documents

CLS40083

Corning® microarray slide mailers/storage boxes

25 slide storage box, clear

Synonym(s):

Corning® 40083

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

manufacturer/tradename

Corning 40083

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

The plastic containers in which UltraGAP Coated Slides are shipped also function as storage boxes for printed arrays. These containers are available as either 5 slide mailers or 25 slide storage boxes. The trans-orange plastic has low transmittance in the 500 to 700 nm wavelength range which helps protect Cy3 and Cy5 dyes from photobleaching. These rigid plastic containers do not shed particles or outgas volatile chemicals that may contaminate microarray slides.
The Corning 25 slider storage box has a lift off lid which is easy to open and close. The 5 slide mailer has a hinged lid that snaps closed tightly to prevent slides from accidentlly falling out.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service