Skip to Content
Merck
All Photos(1)

Key Documents

CLS3907

Corning® Roller Bottles, CellBIND® Cell Culture Surface

round clear polystyrene, sterile, bottle surface area 850 cm2, cap (Easy Grip), case of 40

Synonym(s):

Corning roller bottles, roller bottles

Sign Into View Organizational & Contract Pricing

Select a Size

40 EA
$885.00

$885.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
40 EA
$885.00

About This Item

UNSPSC Code:
41122105
NACRES:
NB.13

$885.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

cap (Easy Grip)
high-density polyethylene cap
polystyrene bottle
round bottle
round clear polystyrene
wide-mouth bottle

sterility

sterile
sterile

feature

cap
disposable
graduations

packaging

case of 40

manufacturer/tradename

Corning 3907

bottle neck diam.

~45 mm

bottle surface area

850 cm2

bottle working volume

170-255 mL

capacity

225 mL

diam. × H

117 mm × 271 mm

binding type

CellBIND® surface

Looking for similar products? Visit Product Comparison Guide

General description


  • The Corning CellBIND cell culture surface
  • Improves cell attachment leading to increased cell growth and yields
  • Results in more consistent and even cell attachment
  • Helps adapt cells to reduced serum or serum-free conditions
  • Reduces premature cell detachment from confluent cultures
Polystyrene roller bottles are also available with a Corning CellBIND surface treatment that uses a novel microwave plasma process for treating the culture surface. This process improves cell attachment by incorporating significantly more oxygen into the cell culture surface than the traditional plasma treatment, rendering it more hydrophilic and increasing the stability of the surface.

Features and Benefits

  • Easy Grip Caps feature large knurls designed for ergonomic handling
  • Easy Grip Vent Caps are designed for applications requiring consistent gas exchange
  • Bottles are manufactured from virgin polystyrene, caps from high density polyethylene
  • Both resins meet the USP Class VI requirements for plastic containers and closures
  • Treated with a Corning CellBIND surface treatment for enhanced cell attachment
  • One piece seamless construction with printed graduations
  • Printed lot numbers to aid in product traceability
  • Sterilized by g−radiation and certified non-pyrogenic

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service