Skip to Content
Merck
All Photos(1)

Key Documents

CLS3677

Corning® 384 well plates, low volume

non-treated, 100/cs, round bottom black polystyrene wells, non-sterile, lid: no

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

black polystyrene plate
round bottom black polystyrene wells

sterility

non-sterile

feature

lid: no

packaging

case of 100
pack of 25

manufacturer/tradename

Corning 3677

size

384 wells

well volume

35 μL

well working volume

2-25 μL

color

black

suitability

suitable for (immunoassay; liquid-handling procedures)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 384 Well Plates, Low Volume

The 384 multiwell low volume plate dimensions conform to proposed SBS standards. The wells for each microplate have either a square to round design accompanied by a flat bottom. In addition, each well has a raised rim to reduce cross contamination during assay and liquid-handling process.
  • Square well top for easier robotic pipetting
  • Round bottom for better Z factor and minimized trapped air
  • Conical well molded in the shape of a light cone for efficiency
  • Black walls reduce well-to-well crosstalk and autofluorescence during fluorescent assays
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm2)

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service