Skip to Content
Merck
All Photos(2)

Key Documents

CLS354224

Corning® rLaminin-521

from human, pack of 500 μg

Synonym(s):

Corning laminin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116155
NACRES:
NA.28
Pricing and availability is not currently available.

biological source

human

form

liquid

packaging

pack of 500 μg

manufacturer/tradename

Corning 354224

technique(s)

cell culture | mammalian: suitable

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Corning rLaminin-521 (Human) is a heterotrimer composed of a5, ß2, and ?1 chains expressed in a mammalian cell culture system. rLaminin-521 (Human) supports long-term self-renewal of human pluripotent stem cells (hPSC), including embryonic stem cells (hESC) and induced pluripotent stem cells (iPSC) in defined and xeno-free environments. rLaminin-521 provides additional benefits, including ROCK inhibitor independent single cell expansion of PSCs, which improves hPSC culture ease and efficiency.

Features

Recombinant protein
Easy to self-coat
Available in 0.5 mg aliquots for easy handling
Compatible with multiple hPSC culture media and dissociation reagents
Supports undifferentiated expansion of hPSC
Suitable for xeno-free, defined culture environment

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service