CLS352058
Corning® Falcon® Round Bottom Test Tubes
capacity 5 mL, cap, snap, sterile, case of 500
Synonym(s):
culture tube
About This Item
Recommended Products
material
round polystyrene
sterility
sterile
feature
cap snap
packaging
case of 500
manufacturer/tradename
Corning 352058
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service