Skip to Content
Merck
All Photos(1)

Key Documents

CLS33971300

Pyrex® graduated funnel, for 47 mm microfiltration assembly

capacity 300 mL

Synonym(s):

pouring funnel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.43
Pricing and availability is not currently available.

material

glass housing

Agency

suitable for EPA 1613

feature

graduation

packaging

pack of 1

manufacturer/tradename

Corning

capacity

300 mL

funnel capacity

300 mL

neck diam.

33-41 mm

Looking for similar products? Visit Product Comparison Guide

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service