Merck
Skip To
CLS3003
Corning® single use spatulas
Tapered blade/spoon
Synonym(s):
Disposable Spatulas
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
NACRES:
NB.17
UNSPSC Code:
41122403
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
sterility
sterile
manufacturer/tradename
Corning 3003
L
247 mm
General description
Corning single use spatulas have been designed to save time and provide contamination-free samples. Eliminates need to recycle and sterilize reusable spatulas.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Features and Benefits
  • Corning® single-use spatulas are made of polystyrene and are color concentrate blends.
  • Tapered blade/spoon with 247 mm length.
  • Visually Inspected for accurate labeling, product configuration, seal and barrier integrity.
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® single use spatulas
Choose from one of the most recent versions:
Certificates of Analysis (COA)