Skip to Content
Merck
All Photos(1)

Key Documents

CLS215010L

Pyrex® single cavity chromatography reservoir

capacity 10 L

Synonym(s):

storage reservoir

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.

material

Pyrex
colorless

grade

for chromatography

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Corning 2150-10L

capacity

10 L

Looking for similar products? Visit Product Comparison Guide

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service