CLS1015P3L
Corning® reusable beaker with handle and spout
polypropylene, size 3 L
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polypropylene
feature
graduations (50 mL)
manufacturer/tradename
Corning 1015P-3L
H × O.D.
243 mm × 170 mm
size
3 L
Looking for similar products? Visit Product Comparison Guide
General description
The polypropylene reusable beakers are transluscent and printed with blue graduations.
Other Notes
Browse all plastic beakers
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service