Skip to Content
Merck
All Photos(1)

Key Documents

BHL-89

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human lymph (axillary)

packaging

tube of 5 μg 98070109-DNA-5UG
pkg of vial of cells 98070109-1VL

growth mode

Suspension

karyotype

Hyperdiploid, modal no. 47, chromosome 1 trisomy, 15 monosomy, +14q+

morphology

Lymphoblastoid

products

Not specified

receptors

sIgM,CD22,BB1,HLA-ABC,GnRH

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human B-type EBNA negative lymphoma

Cell Line Description

The cell line BHL-89 was established from a 56 year old female patient with chronic lymphoid leukaemia (CLL). The B-cell type cells were shown to express CD9, CD22, CD71,HLA-ABC, BB1, GnRH and surface IgM. Cloning efficiency in soft agar was found to be between 20-60% and population doubling time was 48 hours. BHL-89 cells are Epstein-Barr nuclear antigen (EBNA) negative. They have been shown to be tumourigenic in immunosuppressed mice, sensitive to dibromodulcitol and resistant to the phorbol ester TPA. The cell line was found to be positive for BCL-2 protein and produces small amounts of interferon. Lymphoblastoid cells and floating aggregates. The cell line has been eradicated from mycoplasma at ECACC.

Application

B lymphoma studies, cancer treatments

Culture Medium

RPMI1640 + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 1-5 x 100,000 cells/ml;5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service