Skip to Content
Merck
All Photos(1)

Key Documents

LB83

98062328, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LB83, 98062328

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lymphocytes, Xeroderma pigmentosum, EBV-transformed

Cell Line Description

Established by EBV transformation of mononuclear cells from the peripheral blood of a 15 year old female. The patient has been diagnosed with Xeroderma pigmentosum (XP) complementation group G, showed no skin cancer or neurologic abnormalities and reduced NK activity. XP is a relatively rare recessively inherited disease with DNA repair deficiency. Fibroblasts from the same individual have been designated XP125LO.

Application

DNA repair defect studies, hprt mutation analysis

Culture Medium

RPMI 1640 + 4mM Glutamine + 200 μg/ml Sodium Pyruvate (NaPy) + 15% Heat Inactivated (HI-FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml, dilute every 1-2 days to 3 x 100,000 cells/ml 5%

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service