Skip to Content
Merck
All Photos(1)

Documents

M3 Clone M-3

93120827, mouse skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

M3 Clone M-3, 93120827

biological source

mouse skin

growth mode

Adherent

karyotype

Modal no. 83; hypertetraploid

morphology

Epithelial

products

Melanin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse Cloudman S 91 melanoma

Cell Line Description

Melanin producing cell line, derived from a Cloudman S 91 melanoma induced in a F1 (CxDBA) male mouse. The line is tumourigenic in mice and susceptible to a variety of viruses including herpes simplex, vaccinia, pseudorabies and vesicular stomatitis virus (Indiana virus).

Application

Tumourigenicity studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 15% Horse Serum (HS) + 2.5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service