Skip to Content
Merck
All Photos(1)

Key Documents

AFRC MAC 159

Reactivity: Anti-Isopentenyladenosine (2iPA), plant, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 93090999

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Rat Hybridoma

packaging

tube of 5 μg 93090999-DNA-5UG
pkg of vial of cells 93090999-1VL

ImmunoDon

F344 rat spleen

Myeloma

NS0

Reactivity

Anti- isopentenyladenosine (2iPA), plant

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

The antibody is directed against isopentenyl adenosine. In addition it reacts with a range of other cytokines and related compounds.

Immunogen

2iPA-BSA conjugate

Application

Quantification of hormones in plant tissue

Culture Medium

DMEM + 2 mM glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service