Skip to Content
Merck
All Photos(1)

Key Documents

NIH 3T3 Cell Line murine

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 93061524, mouse embryo, Fibroblast

Synonym(s):

3T3 Cells, Mouse 3T3 Cells, NIH/3T3 Cells, NIH3T3 Cells, Swiss 3T3 Cells, Swiss3T3 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse embryo

description

Embryonic fibroblast

packaging

tube of 5 μg 93061524-DNA-5UG
pkg of vial of cells 93061524-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Swiss NIH embryo

Cell Line Description

Established from a NIH Swiss mouse embryo. These cells are sensitive to sarcoma virus focus formation and leukemia virus propogation. Cells have now lost their contact inhibition.

Application

DNA transfection studies
NIH 3T3 Cell Line has been used to:
  • study calcium-mediated actin reset (CaAR) in response to physiological changes[1]
  • study the effects of activation of the transient receptor potential vanilloid 3 channel (TRPV3) on adipocyte differentiation[2]
  • determine the viral titer of the retroviral construct -ST40hTNFα[3]

Culture Medium

DMEM + 2mM Glutamine + 10% Calf Serum (CS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Do not allow culture to become fully confluent; the use of fetal calf serum is not recommended.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Protocols

3D cell culture protocol for generating epidermal human skin tissue using primary human keratinocytes, dermal fibroblasts, and collagen-coated transwell inserts.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service