Skip to Content
Merck
All Photos(1)

Key Documents

Hepa 1-6

92110305, mouse liver, Not specified

Synonym(s):

Hepa 1-6 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

Hepa 1-6, 92110305

biological source

mouse liver

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Albumin, alpha-foetoprotein, alpha 1-antitrypsin, amylase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse hepatoma

Cell Line Description

Derived from BW7756 tumour in a C57L mouse. Secretes several liver products including albumin, α-fetoprotein, α 1-antitrypin and amylase.

Application

Hepa 1-6 has been used to study the biodistribution of polymer hydrogel capsules for the delivery of therapeutics.[1] It has also been used to evaluate the potential of interferon regulatory factor-1 (IRF-1) for cytokine gene therapy of hepatocellular carcinoma (HCC).[2]

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service