The SKU 92090408-DNA-5UG is 5 micrograms of the extracted and purified DNA from this cell line. The 92090408-1VL is the cryopreserved cell line available for culture. The frozen cell lines are packaged as 2 x 10^6 cells per vial.
455
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 92090408, Reactivity: Anti-Epidermal Growth Factor (EGF) Receptor
About This Item
Recommended Products
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 92090408-DNA-5UG
pkg of vial of cells 92090408-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
NS1
Reactivity
Anti-Epidermal Growth Factor (EGF) Receptor
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Cell Line Description
Immunogen
Culture Medium
Subculture Routine
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Quelle est la différence entre ces deux références : 92090408-DNA-5uG et 92090408 1 vial
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service