Skip to Content
Merck
All Photos(1)

Documents

COR-L88

92031917, human lung, Not specified

Synonym(s):

COR-L88 Cell Line, Human COR-L88, Plasmid COR-L88

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COR-L88, 92031917

biological source

human lung

growth mode

Semi-adherent

karyotype

Not specified

morphology

Not specified

products

High levels of L-dopa decarboxylase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian lung small cell carcinoma

Cell Line Description

Derived from the pleural effusion of a 55-year-old Caucasian male.

Application

Studies on lung cancer

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 11
D16S539: 11,12
D5S818: 13
D7S820: 9
THO1: 7
TPOX: 8,10
vWA: 14,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells attach loosely to surfaces and can be aspirated or shaken off.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service