Skip to Content
Merck
All Photos(1)

Key Documents

3T3-MSV

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

mouse embryo

packaging

tube of 5 μg 90092701-DNA-5UG
pkg of vial of cells 90092701-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Growth factor

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

General description

Product has lost contact inhibition

Cell Line Origin

Mouse Swiss 3T3 embryo, Moloney-MSV transformed

Cell Line Description

This cell line is of Swiss 3T3 origin, transformed by the Moloney murine sarcoma virus. It is a non-producer of the virus particles. In serum-free media the line will release a growth factor which can be used to stimulate quiescent cells to grow in monolayer as well as in soft agar.

Application

Cell growth studies

Culture Medium

DMEM + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:6 to 1:8 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Care is required during subculture as cells have a tendency to easily detach from the tissue culture flask.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service