Skip to Content
Merck
All Photos(1)

Key Documents

NCTC 2071

90050803, mouse connective tissue, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

NCTC 2071, 90050803

biological source

mouse connective tissue

growth mode

Adherent

karyotype

Modal no. 63

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C3H/An connective tissue

Cell Line Description

Derivative of NCTC clone 929 and was established in chemically defined medium (NCTC 109). It was subsequently adapted to cysteine-free medium (NCTC 135). The cells have been used in nutritional, immunological and toxicological studies; nutritional, metabolic and enzyme studies; preservation and storage of viable tissue; fluid suspension culture methods and replicate culture techniques and quantitative cell culture methods.

Application

Nutritional, immunological and toxicological studies

Culture Medium

NCTC 135 100% + 2mM Glutamine.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:5 i.e. seeding at 5x10,000 to 2x100,000 cells/cm2 by scraping off the cells and resuspending into new flasks; 5% CO2; 37°C. Freeze in 9% glycerol/91% FBS.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service