Skip to Content
Merck
All Photos(1)

Documents

153BR

90011808, human skin, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

153BR, 90011808

biological source

human skin

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human skin fibroblast

Cell Line Description

Established from a biopsy of a normal 56 year old male. The cells have been used in radiation survival studies. EBV-transformed lymphocytes designated LB185 (Sigma Catalogue number. 98062318) have been isolated from the same individual. This has been confirmed by STR profiling at ECACC.

Application

DNA repair studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 11
D16S539: 9
D5S818: 10,13
D7S820: 11
THO1: 8,9
TPOX: 8,10
vWA: 17,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service