Skip to Content
Merck
All Photos(1)

Documents

CCRF-SB

89090405, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CCRF-SB, 89090405

biological source

human blood

growth mode

Suspension

karyotype

2n = 46

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian acute lymphoblastic leukaemia

Cell Line Description

A B lymphoblastoid line obtained from the peripheral blood of an 11 year old boy with acute lymphoblastic leukaemia. Appear free of virus particles by electron microscopy. Do not synthesise immunoglobulins. Grown in continuous spinner culture for 6 months. HLA profile: A1, A2, B12, B17, Cw2.

Application

HLA profile studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3x10,000 to 1x100,000 cells/ml; 5% CO2; 37°C. Cultures can tolerate high densities i.e. 1-2x1,000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service