Skip to Content
Merck
All Photos(1)

Documents

DDT1-MF2

89051703, hamster testis, Myoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

DDT1-MF2, 89051703

biological source

hamster testis

growth mode

Adherent

karyotype

Not specified

morphology

Myoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Syrian Hamster leiomyosarcoma (Mesocricetus Auratus)

Cell Line Description

A smooth muscle cell line derived from a leiomyosarcoma of the ductus deferens of a Syrian hamster. Growth is stimulated by androgens and inhibited by glucocorticoids.

Application

Tumourigenicity and hormone studies

Culture Medium

DMEM + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Dilute cultures 1:20 dislodging cells by mechanical agitation (0.25% trypsin may be used if necessary); 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service