Skip to Content
Merck
All Photos(1)

Key Documents

LoVo Cell Line human

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 87060101, human colon (supraclavicular lymph node metastasis), Epithelial

Synonym(s):

LoVo Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human colon (supraclavicular lymph node metastasis)

packaging

tube of 5 μg 87060101-DNA-5UG
pkg of vial of cells 87060101-1VL

growth mode

Adherent

karyotype

Modal no. 49, (2n=46)

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon adenocarcinoma

Cell Line Description

Derived from a metastatic tumour in the left supraclavicular region of a 56 year old male with adenocarcinoma of the colon. The cells produce carcino embryonic antigen (CEA).

Application

Human LoVo cell line is part of the three-dimensional (3D) heterotypic spheroid model developed to assess cancer immunotherapy agents in vitro. It has been used to test the in vitro cytotoxic property and anti-proliferative property of C. lechleri stem bark essential oil.[1][2]
Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,13,14
D13S317: 8,11
D16S539: 9,12
D5S818: 11,13
D7S820: 10,11
THO1: 9.3
TPOX: 8,9
vWA: 17,18

Culture Medium

Ham′s F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service