Skip to Content
Merck
All Photos(1)

Documents

BC3H1

86093001, mouse brain, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BC3H1, 86093001

biological source

mouse brain

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Acetylcholine

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse brain tumour

Cell Line Description

Established in 1974 from a mouse brain neoplasm tumour induced by nitrosoethylurea, possess many properties characteristic of smooth muscle. Since then it has been suggested that BC3H1 is a skeletal line of ectodermal origin that is defective for commitment to terminal differentiation. The two distinct morphologies typical of this cell line are large spheroid cells and smooth-muscle-like cells. The fusiform is the more common.

Application

Acetylcholine receptor studies.

Culture Medium

DMEM + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Spilt confluent cultures 1:3 to 1:10 i.e. seeding at 1-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service