Mycl-3C7
85102203, Reactivity: Anti-c-Myc gene product, human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 85102203-DNA-5UG
pkg of vial of cells 85102203-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
Sp2/0
Reactivity
Anti-c-Myc gene product, human
technique(s)
cell culture | mammalian: suitable
isotype
IgG1, kappa
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse Sp2/0 x Mouse BALB/c spleen
Cell Line Description
The antibody recognises the human p62c-myc gene product. It is specific for the peptide D sequence and cross reacts with the p64/66 gene product of the mouse c-myc gene.
Immunogen
A synthetic peptide, corresponding to residues 171-188 (Peptide D) of human p62c-myc sequence, conjugated to KLH
Culture Medium
RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service