A clonal derivative of an SV40 transformed permanent line of MRC-5. Selection was made using X-ray induction in the presence of 2μg/ml thioguanine. The cells are HAT sensitive.
Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)