Skip to Content
Merck
All Photos(1)

Key Documents

JB1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

packaging

tube of 5 μg 01091808-DNA-5UG
pkg of vial of cells 01091808-1VL

ImmunoDon

BALB/c B27 female transgenic mouse

Myeloma

PS-NS1/Ag 4.1

Reactivity

Anti- aggregated HFE protein, Human

technique(s)

cell culture | mammalian: suitable

isotype

IgG3

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Anti-recombinant HFE Protein hybridoma

Cell Line Description

To produce this hybridoma mice transgenic HLA-B27 and human β-2 microglobulin were immunized with a preparation of HFE made in recombinant bacteria, refolded with human β-2 microglobulin. Characterisation beyond data in B.J. Haematol 103, 931, 1998 shows that JB1 reacts predominantly with aggregated denatured HFE produced in bacteria, but does not bind folded recombinant HFE produced in CHO cells. In comparison with polyclonal sera raised to the C terminus or the α 3 region of HFE JB1 stains frozen sections of the tissues well, but reacts very weakly with cells transfected with the HFE gene in vitro (usually only visible by confocal microscopy). JB1 does not stain paraffin fixed sections and will not work in western blots.

Immunogen

Bacterial recombinant HFE protein

Culture Medium

RPMI 1640, 10% FBS, 1% HAT

Subculture Routine

Maintain cells at 3-5x100,000 cells/ml which is usually a split ratio of 1:2. Saturation density os 5 x 100,000 to 1 x 1000,000.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service