C9274
Cytidine 5′-triphosphate disodium salt
~95%
Synonym(s):
CTP
About This Item
Recommended Products
Assay
~95%
storage temp.
−20°C
SMILES string
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O
InChI
1S/C9H16N3O14P3.2Na/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H2,10,11,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1
InChI key
NFQMDTRPCFJJND-WFIJOQBCSA-L
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Preparation Note
comparable product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service