Skip to Content
Merck
All Photos(1)

Key Documents

C8624

Sigma-Aldrich

COG 133 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Ac- LRVRLASHLRKLRKRLL-NH2, Ac-Leu-Arg-Val-Arg-Leu-Ala-Ser-His-Leu- Arg-Lys-Leu-Arg-Lys-Arg-Leu-Leu-NH2, COG133, N-acetyl-L-leucyl-L-arginyl-L-valyl-L-arginyl-L-leucyl-L-alanyl-L-seryl-L-histidyl-L-leucyl-L-arginyl-L-lysyl-L-leucyl-L-arginyl-L-lysyl-L-arginyl-L-leucyl-L-Leucinamide, apoE(133-149)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$344.00

$344.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$344.00

About This Item

Empirical Formula (Hill Notation):
C97H181N37O19 · xC2HF3O2
CAS Number:
Molecular Weight:
2169.71 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

$344.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

color

white

solubility

H2O: >1 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc1cnc[nH]1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)ON

InChI

1S/C97H181N37O20.C2HF3O2/c1-50(2)40-67(119-58(16)136)85(146)125-66(32-25-39-116-97(108)109)83(144)134-75(56(13)14)91(152)126-65(31-24-38-115-96(106)107)82(143)127-68(41-51(3)4)84(145)118-57(15)76(137)133-74(48-135)90(151)131-72(46-59-47-111-49-117-59)89(150)130-70(43-53(7)8)87(148)124-63(29-22-36-113-94(102)103)79(140)121-61(27-18-20-34-99)80(141)128-69(42-52(5)6)86(147)123-62(28-21-35-112-93(100)101)78(139)120-60(26-17-19-33-98)77(138)122-64(30-23-37-114-95(104)105)81(142)129-71(44-54(9)10)88(149)132-73(45-55(11)12)92(153)154-110;3-2(4,5)1(6)7/h47,49-57,60-75,135H,17-46,48,98-99,110H2,1-16H3,(H,111,117)(H,118,145)(H,119,136)(H,120,139)(H,121,140)(H,122,138)(H,123,147)(H,124,148)(H,125,146)(H,126,152)(H,127,143)(H,128,141)(H,129,142)(H,130,150)(H,131,151)(H,132,149)(H,133,137)(H,134,144)(H4,100,101,112)(H4,102,103,113)(H4,104,105,114)(H4,106,107,115)(H4,108,109,116);(H,6,7)/t57-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,70-,71-,72-,73-,74-,75-;/m0./s1

InChI key

SIAUOBGYBRMVPH-BGJGLIJFSA-N

Biochem/physiol Actions

COG 133 is an apoE protein fragment that blocks neurodegeneration and restores cognitive functions.
COG 1333 or apoE(133-149) is a fragment of Apolipoprotein E (APOE) that mimics the functional anti-inflammatory and neuroprotective effects of the intact apoE protein. It was suggested that neuroprotective functions of the mimetic results from binding to the low-density lipoprotein receptor-related protein (LRP) or LDL receptors, and such binding results in inhibition of the N-methyl-D-aspartate (NMDA) receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fernando Lopes et al.
Arthritis & rheumatology (Hoboken, N.J.), 68(2), 542-552 (2015-09-01)
Activation of antiinflammatory cholinergic (vagal) pathways can reduce inflammation, and in vitro studies support a pivotal role of α7 nicotinic acetylcholine receptors (α7-nAChR), macrophages, and T cells in these events. The aim of this study was to assess α7-nAChR agonists

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service