Skip to Content
Merck

Skip To

C8605

Dihydroceramide C8

98%, waxy solid

Synonym(s):

D-erythro-N-Octanoyldihydrosphingosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 mg
Check Cart for Availability
$594.00

About This Item

Empirical Formula (Hill Notation):
C26H53NO3
Molecular Weight:
427.70
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Assay:
98%
Form:
waxy solid
Quality level:

$594.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

98%

form

waxy solid

color

white

solubility

ethanol: 10 mg/mL, DMF: 25 mg/mL, DMSO: 5 mg/mL

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI

1S/C26H53NO3/c1-3-5-7-9-10-11-12-13-14-15-16-18-19-21-25(29)24(23-28)27-26(30)22-20-17-8-6-4-2/h24-25,28-29H,3-23H2,1-2H3,(H,27,30)/t24-,25+/m0/s1

InChI key

LGOFBZUQIUVJFS-LOSJGSFVSA-N

Biochem/physiol Actions

Inactive form of C8 ceramide for use as a negative control.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7980C8480376650
form

waxy solid

form

solid

form

solid

form

waxy solid

assay

98%

assay

≥97% (TLC)

assay

98% (TLC)

assay

≥98% (TLC)

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

ethanol: 10 mg/mL, DMSO: 5 mg/mL, DMF: 25 mg/mL

solubility

DMSO: 5 mg/mL (Dissolve in hot DMSO, then cool to RT.), ethanol: 5 mg/mL

solubility

DMSO: 25 mg/mL, ethanol: 25 mg/mL

solubility

ethanol: 25 mg/mL, DMSO: soluble

color

white

color

white

color

white

color

white


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Y W Hsu et al.
Journal of immunology (Baltimore, Md. : 1950), 166(9), 5388-5397 (2001-04-21)
The goal of this study was to elucidate whether triggering the sphingomyelin pathway modulates LPS-initiated responses. For this purpose we investigated the effects of N-acetylsphingosine (C(2)-ceramide) on LPS-induced production of NO and PGE(2) in murine RAW 264.7 macrophages and explored
Mike Lange et al.
Cell reports. Medicine, 2(10), 100407-100407 (2021-11-11)
Obesity, characterized by expansion and metabolic dysregulation of white adipose tissue (WAT), has reached pandemic proportions and acts as a primer for a wide range of metabolic disorders. Remodeling of WAT lipidome in obesity and associated comorbidities can explain disease



Global Trade Item Number

SKUGTIN
C8605-25MG04061833261200
C8605-5MG04061833261217

Questions

Reviews

No rating value

Active Filters