Skip to Content
Merck
All Photos(1)

Key Documents

C8230

Sigma-Aldrich

Dihydroceramide C6

solid

Synonym(s):

N-Hexanoyldihydrosphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00

$166.00


Estimated to ship onMarch 29, 2025



Select a Size

Change View
5 MG
$166.00

About This Item

Empirical Formula (Hill Notation):
C24H49NO3
Molecular Weight:
399.65
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

$166.00


Estimated to ship onMarch 29, 2025


form

solid

Quality Level

color

white

solubility

DMF: soluble
DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI

1S/C24H49NO3/c1-3-5-7-8-9-10-11-12-13-14-15-16-18-19-23(27)22(21-26)25-24(28)20-17-6-4-2/h22-23,26-27H,3-21H2,1-2H3,(H,25,28)/t22-,23+/m0/s1

InChI key

VUMHYWBWYSPQMM-XZOQPEGZSA-N

Biochem/physiol Actions

Inactive form of C6 ceramide for use as a negative control.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kitatani et al.
Archives of biochemistry and biophysics, 382(2), 296-302 (2000-11-09)
The effect of ceramide on Ca2+-dependent translocation of cytosolic phospholipase A2 (cPLA2) to membranes was studied. Pretreatment of platelets with sphingomyelinase or C6-ceramide (N-hexanoylsphingosine) led to apparent enhancement of Ca2+-ionophore A23187-stimulated arachidonic acid release but did not affect the cytosolic
F J Sánchez-Gómez et al.
Allergy, 72(3), 385-396 (2016-06-21)
Allergic reactions to β-lactams are among the most frequent causes of drug allergy and constitute an important clinical problem. Drug covalent binding to endogenous proteins (haptenation) is thought to be required for activation of the immune system. Nevertheless, neither the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service