Skip to Content
Merck
All Photos(3)

Key Documents

C8210

Sigma-Aldrich

CHES

BioXtra, ≥99.0% (titration)

Synonym(s):

2-(Cyclohexylamino)ethanesulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$81.50
100 G
$330.00

$81.50

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$81.50
100 G
$330.00

About This Item

Empirical Formula (Hill Notation):
C8H17NO3S
CAS Number:
Molecular Weight:
207.29
Beilstein:
2967601
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$81.50

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

product line

BioXtra

Assay

≥99.0% (titration)

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

pH

3.0-5.0 (0.5 M in H2O)

useful pH range

8.6-10.0

pKa (25 °C)

9.3

solubility

H2O: 0.5 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.0005%

absorption

≤0.05 at 280 in H2O at 0.5 M
≤0.08 at 260 in H2O at 0.5 M

application(s)

diagnostic assay manufacturing

SMILES string

OS(=O)(=O)CCNC1CCCCC1

InChI

1S/C8H17NO3S/c10-13(11,12)7-6-9-8-4-2-1-3-5-8/h8-9H,1-7H2,(H,10,11,12)

InChI key

MKWKNSIESPFAQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CHES (2-(Cyclohexylamino) ethane sulfonic acid) is a derivative of amino acid taurine. It serves as a standard physiological buffer in the pH range of 8.5-9.5.[1]

Application

CHES may be used:
  • for pH adjustments and as a assay buffer in catechol oxidase activity measurements(32)
  • as a buffer in electron paramagnetic resonance (EPR) spectroscopy and kinetic measurements(33)
  • as a buffer for soil solution equivalent (SSE) yeast dextrose medium for screening pH optima for growth of Aridibacter nitratireducens A24_SHP-5_238T(34)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Buffers for the physiological pH range: Acidic dissociation constants of zwitterionic compounds (ACES and CHES) in water from 5 to 55? C
Roy RN, et al.
Journal of Chemical and Engineering Data, 42(1), 41-44 (1997)
Jun Yang et al.
Langmuir : the ACS journal of surfaces and colloids, 24(18), 10265-10272 (2008-08-20)
A universal nitric oxide (NO) generating surface is assembled via Layer-by-Layer (LbL) deposition of sodium alginate (Alg) and organoselenium modified polyethyleneimine (SePEI) on quartz and polymeric substrates. The immobilized SePEI species is capable of catalytically decomposing S-nitrosothiol species (RSNO) to
Scott A Sarver et al.
Journal of chromatography. A, 1229, 268-273 (2012-02-11)
Several glycosphingolipids were labeled with the fluorphore Bodipy-Fl and analyzed using capillary electrophoresis with laser-induced fluorescence detection. GM1-, LacCer-, and Cer-Bodipy-Fl were prepared through acylation using the N-hydroxysuccinimide ester of Bodipy-Fl. Several other glycosphingolipids including GT1a-, GD1a-, GM2-, GM3-, GD3-
Luis Felipe Vallejo et al.
Biotechnology and bioengineering, 85(6), 601-609 (2004-02-18)
The human gene encoding the mature form of bone morphogenetic protein-2 (hBMP-2), a dimeric disulfide-bonded protein of the cystine knot growth factor family, was expressed in recombinant Escherichia coli using a temperature-inducible expression system. The recombinant protein was produced in
Takashi Miyado et al.
Journal of chromatography. A, 1206(1), 41-44 (2008-08-12)
In order to develop a high-throughput assay for nitric oxide metabolites, nitrite (NO2-) and nitrate (NO3-), in biological fluids, we have investigated the simultaneous determination of them using an electrophoretic lab-on-a-chip (microchip capillary electrophoresis, MCE) technique. In this study, in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service