Skip to Content
Merck

Skip To

C7482

Caspase 6 human

recombinant, expressed in E. coli, >90% (SDS-PAGE), lyophilized powder, 13,000 units/mg protein

Synonym(s):

Mch2

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Specific activity:
13,000 units/mg protein
Assay:
>90% (SDS-PAGE)
Recombinant:
expressed in E. coli


recombinant

expressed in E. coli

Quality Level

assay

>90% (SDS-PAGE)

form

lyophilized powder

specific activity

13,000 units/mg protein

mol wt

~30 kDa

solubility

PBS: soluble

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

Cleaves the substrates consisting of consensus sequence VEID.
Spontaneously undergoes autoprocessing to yield the subunits associated to form the active enzyme.
Useful in screening caspase inhibitors, studying enzyme regulation, determining specificity of caspase substrates, or as a positive control in caspase activity assays.

Physical form

Contains 0.052% ammonium sulfate, 0.158% Tris−HCl, 0.76% sodium chloride.

Other Notes

One unit will hydrolyze 1 nmol of the caspase substrate VEID-pNA to VEID and p-nitroaniline per hour at pH 7.2 at 37 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
218783C1099C6357
assay

>90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

-

specific activity

13,000 units/mg protein

specific activity

≥25,000 units/mg protein, ≥50,000 units/mL

specific activity

≥1,000 units/mg protein

specific activity

5,000 units/mg protein

form

lyophilized powder

form

liquid

form

buffered aqueous solution

form

solution

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli (C-terminal histidine tagged)

recombinant

expressed in E. coli

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ana A Baburamani et al.
Developmental neuroscience, 37(4-5), 321-337 (2015-04-01)
Apoptotic mechanisms are centre stage for the development of injury in the immature brain, and caspases have been shown to play a pivotal role during brain development and in response to injury. The inhibition of caspases using broad-spectrum agents such



Questions

Reviews

No rating value

Active Filters