Skip to Content
Merck
All Photos(1)

Key Documents

C7370

Sigma-Aldrich

CAMKK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAMKKβ, KIAA0787, MGC15254

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

9-13 nmol/min·mg

mol wt

~88 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CAMKK2(10645)

Biochem/physiol Actions

CAMKK2 (CAMKKb) is a member of the CAMKK family. It is broadly distributed in tissues with highest levels in brain, thymus, spleen, and testis. CAMKK2 undergoes intramolecular autophosphorylation, is regulated by Ca2+/calmodulin and phosphorylates CAMKI and CAMKIV on Thr177 and Thr200, respectively. CAMKK2 activates both CAMKI and CAMKIV when coexpressed in Jurkat T cells. CAMKK2 also phosphorylates and regulates the activity of AMPK, which is an important regulator of cellular metabolism in response to metabolic stress.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian York et al.
Scientific reports, 7(1), 11793-11793 (2017-09-20)
Binding of calcium to its intracellular receptor calmodulin (CaM) activates a family of Ca
Rebecca L Hurley et al.
The Journal of biological chemistry, 280(32), 29060-29066 (2005-06-28)
The AMP-activated protein kinase (AMPK) is an important regulator of cellular metabolism in response to metabolic stress and to other regulatory signals. AMPK activity is absolutely dependent upon phosphorylation of AMPKalphaThr-172 in its activation loop by one or more AMPK
K A Anderson et al.
The Journal of biological chemistry, 273(48), 31880-31889 (1998-11-21)
Ca2+/calmodulin-dependent protein kinases I and IV (CaMKI and CaMKIV, respectively) require phosphorylation on an equivalent single Thr in the activation loop of subdomain VIII for maximal activity. Two distinct CaMKI/IV kinases, CaMKKalpha and CaMKKbeta, were purified from rat brain and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service