Skip to Content
Merck
All Photos(2)

Key Documents

C7210

Sigma-Aldrich

N-Z-Amine® HD

Synonym(s):

Peptone from casein

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$222.00
500 G
$404.00

$222.00


Estimated to ship onMay 12, 2025



Select a Size

Change View
250 G
$222.00
500 G
$404.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

$222.00


Estimated to ship onMay 12, 2025


biological source

Porcine
bovine

Quality Level

description

Amino nitrogen ≥ 8%
Total nigrogen ≥11%

form

powder

technique(s)

microbiological culture: suitable

Looking for similar products? Visit Product Comparison Guide

Application

N-Z-Amine® has been used in culture mediums to grow and produce enterotoxins by Staphylococcus.

Quality

Preparation has a high level of free amino acids.

Legal Information

N-Z-Amine is a registered trademark of Kerry Group

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T N Morita et al.
Applied and environmental microbiology, 38(1), 39-42 (1979-07-01)
From comparisons of 4% N-Z Amine NAK made with distilled water, naturally hard water, and synthetic salt solutions, it appeared that magnesium and, to a lesser extent, iron were limiting factors in the production of staphylococcal enterotoxins B and C
J F Metzger et al.
Applied microbiology, 25(5), 770-773 (1973-05-01)
Release of Staphylococcus aureus enterotoxin B (SEB) into the culture medium was initiated during the mid-log phase of growth. A medium consisting of 4% N-Z Amine A (Sheffield), 0.2% dextrose, and 1% yeast extract supported maximum production of SEB. Although
C Genigeorgis et al.
Applied microbiology, 21(5), 862-866 (1971-05-01)
Growth and production of enterotoxin C by Staphylococcus aureus strain 137 in 3% + 3% protein hydrolysate powder N-Z Amine NAK broths with 0 to 12% NaCl and an initial pH of 4.00 to 9.83 were studied during an 8-day

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service