Skip to Content
Merck
All Photos(1)

Key Documents

C7089

Sigma-Aldrich

Monoclonal Anti-Cdk7/CAK antibody produced in mouse

clone MO-1.1, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

MO-1.1, monoclonal

mol wt

antigen 40 kDa

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1:50,000 using a cultured human cell line extract

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK7(1022)

General description

Cell division protein kinase 7 is an enzymes encoded by the CDK7 gene in humans. The protein belongs to cyclin-dependent protein kinase (CDK) family. CAK is a CDK-cyclin complex implicated in the control of multiple cell cycle transitions. The structure of CDK7 facilitates the inhibitors with differential specificity for the different CDKs and can be used in different biological processes.

Specificity

The antibody reacts specifically with Cdk7 (40 kDa catalytic subunit of Cdk activating kinase).

Immunogen

recombinant human Cdk7 protein.

Application

Monoclonal Anti-Cdk7/CAK antibody produced in mouse is suitable for the following applications:
  • Immunocytochemistry
  • Immunohistochemistry (frozen sections)
  • Immunoprecipitation
  • Microarray
  • Western blotting at a dilution of 1:50,000 using a cultured human cell line extract

Biochem/physiol Actions

Cyclin-dependent kinase (CDK) Activating Kinase (CAK) is involved in activation and phosphorylation of CDK1, CDK2, CDK4 and CDK6 and regulation of the cell cycle. It is required for cell cycle progression and it could be a target for cancer therapy. CAK plays a central role in regulation of messenger RNA synthesis by either inhibition of RNA polymerase II-catalyzed transcription or stimulation of transcription through association with basal transcription repair factor IIH. Phosphorylation by the CDK-activating kinase (CAK) is required for the activation of cyclin-dependent kinases.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel Rovnak et al.
Journal of virology, 86(10), 5742-5751 (2012-03-02)
Alterations in the functional levels of cyclin-dependent kinase-8 (CDK8) or its partner, cyclin C, have been clearly associated with cancers, including colon cancer, melanoma, and osteosarcoma. Walleye dermal sarcoma virus encodes a retroviral cyclin (RV-cyclin) that localizes to interchromatin granule
Stéphane Larochelle et al.
Methods in molecular biology (Clifton, N.J.), 296, 279-290 (2004-12-04)
All cyclin-dependent kinases (CDKs) involved in eukaryotic cell cycle control require phosphorylation at a conserved threonine (or serine) residue within the activation- or T-loop to attain full enzymatic activity. The enzyme responsible for this activating phosphorylation, the CDK-activating kinase (CAK)
R P Fisher et al.
Cell, 78(4), 713-724 (1994-08-26)
Phosphorylation by the CDK-activating kinase (CAK) is a required step in the activation of cyclin-dependent kinases. We have purified CAK from mammalian cells; the enzyme comprises two major polypeptides of 42 and 37 kDa. Protein sequencing indicates that the 42
D A Bochar et al.
The Journal of biological chemistry, 274(19), 13162-13166 (1999-05-01)
Cyclin-dependent kinase 7 (CDK7) can be isolated as a subunit of a trimeric kinase complex functional in activation of the mitotic promoting factor. In this study, we demonstrate that the trimeric cdk-activating kinase (CAK) acts as a transcriptional repressor of
Graziano Lolli et al.
Cell cycle (Georgetown, Tex.), 4(4), 572-577 (2005-05-07)
The Cyclin-dependent kinase (CDK) Activating Kinase (CAK) is responsible for the activating phosphorylation of CDK1, CDK2, CDK4 and CDK6 and regulation of the cell cycle. The kinase is composed of three subunits: CDK7, Cyclin H and MAT1 (ménage a trois).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service